Learn More
Medchemexpress LLC Arcyriaflavin A | 118458-54-1 | MFCD03452662 | 99.8% | 325.32 g·mol⁻¹ | C20H11N3O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1033821MG
Encompass_Preferred
Arcyriaflavin A is a fungal metabolite isolated from Nocardiopsis species. It acts as a kinase inhibitor and biochemical probe, showing activity against CDK4 and CaMKII, and is intended for laboratory research use.
  • Inhibits CDK4 (IC50 = 140 nM) and CaMKII (IC50 = 25 nM).
  • High purity: 99.8%.
  • Molecular formula C20H11N3O2; molecular weight 325.32 g·mol⁻¹.
  • Supplied as a small research pack suitable for biochemical and enzymatic assays.
Catalog No. 50-002-44720