Learn More
Cayman Chemical ARCYRIAFLAVIN A 5MG
SDP
Supplier:  Cayman Chemical 135165 MG
Encompass_Preferred
A CDK4 and CaMKII inhibitor (IC50s = 140 and 25 nM, respectively); selective for CDK4 and CaMKII over PKA and PKC (IC50s = >2 and >100 µM, respectively); inhibits replication of HCMV in vitro (IC50 = 200 nM); inhibits proliferation of HCT116 and NCI H460 carcinoma cells in vitro (IC50s = 850 and 590 nM, respectively); inhibits proliferation and induces apoptosis of ECSCs
Catalog No. 50-295-5649