Learn More
Aviva Systems Biology Corp Ard1b Antibody - middle region (ARP48601_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48601_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ard1b. It was validated on Western Blot. . Alternate names: Naa11, Ard1bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-370