Learn More
Medchemexpress LLC Arecaidine hydrobromide | 6013-57-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN2368B5MG
Encompass_Preferred
Arecaidine hydrobromide is a pyridine alkaloid known for its role as a potent GABA uptake inhibitor. It also acts as a substrate for H+-coupled amino acid transporter 1 (PAT1, SLC36A1) and can competitively inhibit L-proline uptake.
  • Potent GABA uptake inhibitor
  • Substrate of H+-coupled amino acid transporter 1 (PAT1, SLC36A1)
  • Competitively inhibits L-proline uptake
  • White to off-white solid appearance
  • Intended for research use only
Catalog No. 50-004-20774