Learn More
Medchemexpress LLC Arecaidine hydrochloride | 6018-28-6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN2368A5MG
Encompass_Preferred
Arecaidine hydrochloride is a pyridine alkaloid that acts as a potent GABA uptake inhibitor. It is also identified as a substrate of H+-coupled amino acid transporter 1 (PAT1, SLC36A1) and has been shown to competitively inhibit L-proline uptake.
  • Acts as a potent GABA uptake inhibitor
  • Functions as a substrate of H+-coupled amino acid transporter 1 (PAT1, SLC36A1)
  • Competitively inhibits L-proline uptake
Catalog No. 50-004-20542