Learn More
Selleck Chemical LLC Arenobufagin S0943-5MG
SDP
Supplier:  Selleck Chemical LLC S09435MG
Encompass_Preferred
Arenobufagin is a natural bufadienolide from toad venom and induces apoptosis and autophagy in human hepatocellular carcinoma cells through inhibition of PI3K/Akt/mTOR pathway
Catalog No. 50-000-42590