Learn More
Chemglass Life Sciences AREX-6 CONN.PRO AREX-6 with probe, with Vertical Support Rod, Boss Head, and Support/Holding Rod, 115V/60hz Cloud-Enabled Hot Plate Stirrer with Timer
SDP
Supplier:  Chemglass Life Sciences CG1997W50
Encompass_Preferred
Velp Wifi enabled 135mm round top DIGITAL hot plate stirrer system with built in timer. Hotplates feature many exclusive features and improved safety enhancements. The 135mm diameter heater top is scratch proof and chemically resistant. The integrated TIMER provides an additional layer of safety ensuring the hotplate heater is "off" after the elapsed time (settable from 1min to 99h: 59min). TRU-RPM stirring provides very accurate mixing speeds through the entire RPM range from 30 to 1700 rpm. The back corner of the housing has an M8 female thread to accept a vertical support rod. The updated interface, with easy to read, illuminated icons, create a very user-friendly experience. New and improved knobs and buttons assure simple and intuitive control of the hotplate stirrer.
Catalog No. NC1798087