Learn More
Medchemexpress LLC Arformoterol tartrate | 200815-49-2 | 99.6% | C23H30N2O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0010D50MG
Encompass_Preferred
Arformoterol tartrate, the (R,R)-enantiomer of Formoterol, functions as a long-acting β2-adrenergic receptor (β2-AR) agonist with a Kd of 2.9 nM. This compound is utilized in research studies related to chronic obstructive pulmonary disease (COPD).
  • (R,R)-enantiomer of Formoterol
  • Acts as a long-acting β2-adrenergic receptor (β2-AR) agonist
  • Exhibits a Kd of 2.9 nM
  • Induces cAMP accumulation in cultured human bronchial epithelial cells
  • Mitigates Cl2-induced increases of respiratory system resistance and elastance in mice
  • Reverses histamine- and ovalbumin-induced bronchoconstriction in guinea pigs
Catalog No. 50-004-32535