Learn More
Medchemexpress LLC Arg-AMS | 301351-95-1 | 98.0% | 502.51 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1128621MG
Encompass_Preferred
Arg-AMS (compound 24) is a potent nanomolar inhibitor of arginyl tRNA synthetase. It displays tightly bound inhibitory characteristics for the A-domains in non-ribosomal peptide synthetases (NRPS) enzymes and inhibits the Orn-activating domain of GrsB with an IC50 value of 4.6 μM. This product is for research use only and not sold to patients.
  • Potent nanomolar inhibitor.
  • Inhibits arginyl tRNA synthetase.
  • Displays tightly bound inhibitory characteristics for A-domains in non-ribosomal peptide synthetases enzymes.
  • Suitable for research applications.
Catalog No. 50-004-39764