Learn More
Medchemexpress LLC Arg-Arg-AMC acetate | 487.56 (free base) | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP4340A10MG
Encompass_Preferred
Arg-Arg-AMC acetate is the acetate salt form of Arg-Arg-AMC (HY-P4340). It functions as a fluorescent substrate for cathepsin B and is utilized in cathepsin B activity assays.
  • Fluorescent substrate for cathepsin B
  • Used in cathepsin B activity assay
  • Target: Cathepsin
  • Pathway: Metabolic enzyme/protease
Catalog No. 50-003-97930