Learn More
Medchemexpress LLC Arg-Gly-Glu-Ser TFA | 99.5% | 561.47 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP0309A
Encompass_Preferred
Arg-Gly-Glu-Ser TFA is a polypeptide related to RGD that serves as a negative control for Arg-Gly-Asp-Ser (HY-12290). This product is for research use only.
  • Related to RGD polypeptides
  • Negative control for Arg-Gly-Asp-Ser
  • For research applications
  • Does not influence neutrophil and macrophage cell numbers in studies
  • Does not impact protein accumulation in comparative studies
Catalog No. 50-004-26856