Learn More
Medchemexpress LLC Arg-Gly-Tyr-Ser-Leu-Gly | 59587-18-7 | 99.4% | 651.71 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP393510MG
Encompass_Preferred
Arg-Gly-Tyr-Ser-Leu-Gly corresponds to the sequence of residues from 21 through 26 in lysozyme. It can be utilized as a substrate for protein kinase and is phosphorylated at the serine residue by protein kinase.
  • Corresponds to lysozyme residues 21-26
  • Serves as a substrate for protein kinase
  • Phosphorylated at the serine residue by protein kinase
Catalog No. 50-003-97231