Learn More
Medchemexpress LLC Argatroban | 74863-84-6 | MFCD23102419 | 99.8% | 508.63 g/mol | C23H36N6O5S | 10 MG

Supplier: Medchemexpress LLC HYB0375R10MG
Argatroban (Standard) is the analytical standard of argatroban (MD-805), a direct, selective thrombin inhibitor. It is supplied as a solid 10 mg package for research and analytical applications and is intended as a reference material for assay development and quality control (not for human use).
- Analytical reference standard for argatroban assays and method development.
- High reported purity (~99.8%).
- Molecular weight 508.63 g/mol; formula C23H36N6O5S.
- CAS number 74863-84-6 for unambiguous identification.
- Supplied as a 10 mg solid suitable for analytical use.
- Intended for research and analytical applications; not for therapeutic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.