Learn More
Genetex Inc Arginase 1 Monoclonal Antibody [MSVA-511R] HistoMAX™ (GTX04426) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04426
Encompass_Preferred
The Arginase 1 antibody [MSVA-511R] HistoMAX™ (GTX04426) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human liver tissue using GTX04426 Arginase 1 antibody [MSVA-511R] HistoMAX. Strong nuclear and cytoplasmic arginase 1 expression in all hepatocytes in a normal liver.
Catalog No. NC3666926