Learn More
ABclonal Technology Arginase-1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2580820UL
Encompass_Preferred
Enables arginase activity Involved in defense response to protozoan negative regulation of T-helper 2 cell cytokine production and negative regulation of activated T cell proliferation Predicted to be located in several cellular components including extracellular space mitochondrial outer membrane and neuronal cell body Predicted to be active in cytosol Is expressed in several structures including alimentary system central nervous system genitourinary system integumental system and sensory organ Used to study hyperargininemia Human ortholog(s) of this gene implicated in asthma hepatocellular carcinoma and hyperargininemia Orthologous to human ARG1 (arginase 1)
Catalog No. 50-000-09171