Learn More
Medchemexpress LLC Arginase, microorganism (immobilized) | 9000-96-8 | 1 KU
SDP
Supplier:  Medchemexpress LLC HYP31901KU
Encompass_Preferred
Arginase, microorganism immobilized (L-Arginine amidinase) is an enzyme of the urea cycle. Arginase catalyzes the hydrolysis of L-arginine to L-ornithine and urea.
  • Enzyme of the urea cycle
  • Catalyzes the hydrolysis of L-arginine to L-ornithine and urea
  • Available in solid appearance
  • Color ranges from white to light yellow
Catalog No. 50-003-98068