Learn More
Medchemexpress LLC Arginine | 7200-25-1 | 174.20 | 1G
SDP
Supplier:  Medchemexpress LLC HYN04541G
Encompass_Preferred
DL-Arginine is the racemic compound of L-Arginine and D-Arginine. It is an essential amino acid requiring supplementation in traumatic or diseased conditions, playing a role in T cell activation, proliferation, differentiation, and immune cell function. DL-Arginine can serve as a nitrogen source in culture media to support the growth of cells or bacteria.
  • Racemic compound of L-Arginine and D-Arginine
  • Involved in T cell activation, proliferation, and differentiation
  • Affects the function of immune cells
  • Can be used as a nitrogen source in cell and bacteria culture media
Catalog No. 50-003-89783