Learn More
Boston Bioproducts Inc Arginine-HCl Buffer (0.5 M, pH 7.0) - 500ml
Supplier:  Boston Bioproducts Inc BB2505500ML
Encompass
The Arginine-HCl buffer is used to maintain the pH within physiological range, which is essential for processes that demand consistent environments. It is effective in several applications including the purification and precipitation of nucleic acids, as well as in protein crystallization and gel staining during electrophoresis. The buffering capacity of an arginine buffer is influenced by its concentration, while its pH can fluctuate based on the ratio of arginine to arginine hydrochloride.
Catalog No. NC2104292