Learn More
Medchemexpress LLC Arginyl-Glutamine TFA | 2483-17-2 | 99.2% | 644.40 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP4253A1MG
Encompass_Preferred
Arginyl-Glutamine TFA is a dipeptide that can decrease VEGF levels and inhibit retinal neovascularization in a mouse model of oxygen-induced retinopathy. It is intended for research use only.
  • Dipeptide compound
  • Decreases VEGF levels
  • Inhibits retinal neovascularization in mouse models
  • For research use only
Catalog No. 50-003-97657