Learn More
Medchemexpress LLC Arginyl-glutamine TFA | 99.2% | 644.40 | 10 MG

Supplier: Medchemexpress LLC HYP4253A10MG
Arginyl-Glutamine TFA is a dipeptide that can decrease VEGF levels and inhibit retinal neovascularization in a mouse model of oxygen-induced retinopathy. It is for research use only.
- Dipeptide
- Decreases VEGF levels
- Inhibits retinal neovascularization
- Appearance: solid
- Sequence: Arg-Gln
- Sequence shortening: RQ
- Shipping: room temperature in continental US
- Storage (powder): -80°C for 2 years, -20°C for 1 year
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month (sealed storage, away from moisture)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.