Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Arginyl-glycyl-aspartyl-serine | 91037-65-9 | 99.1% | 50 MG
SDP

Supplier:  Medchemexpress LLC HY1229050MG

Encompass_Preferred

Arg-Gly-Asp-Ser (RGDS peptide) is a tetrapeptide integrin-binding sequence used to inhibit integrin receptors and as an analytical standard in cell-adhesion and receptor-ligand assays. It is supplied as a white to off-white solid suitable for biochemical and cell-based research.

  • Tetrapeptide sequence: Arg-Gly-Asp-Ser (RGDS).
  • High purity (about 99.1%).
  • Molecular weight 433.42 g/mol; formula C15H27N7O8.
  • Available as powder or solution (for example, 10 mM in DMSO) in multiple sizes, including 50 MG.
  • Store sealed, away from moisture: powder -80°C (long term) or -20°C (short term).
  • Used in integrin inhibition, cell-adhesion assays, and receptor-ligand studies.

Catalog No. 50-002-31051


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.