Learn More
GENERAL BIOPHYSICS LLC Argon Gas Analyzer Model B
Supplier:  GENERAL BIOPHYSICS LLC GAARO2B
Encompass
Thermoconductivity Gas Analyzer for Argon (In-line Monitoring)Application: Real-time, in-line concentration monitoring for xenon gas.Accuracy: ±1%.
Catalog No. NC3361680