Learn More
Medchemexpress LLC Argonaute-2 inhibitor BCI-137 | 112170-24-8 | MFCD05148320 | 98.9% | 313.29 | C11H11N3O6S | 10 MG

Supplier: Medchemexpress LLC HY11902410MG
BCI-137 is a small-molecule competitive inhibitor of Argonaute 2 (AGO2) that binds to residues in the MID domain. It is supplied for laboratory research and is provided as a solid or as a DMSO solution.
- Competitive AGO2 inhibition with a reported IC50 of 342 μM.
- Molecular weight 313.29 and formula C11H11N3O6S.
- Available as a solid (typical 10 mg) or as a 10 mM solution in DMSO.
- Storage stability: powder stable at -20°C for extended periods.
- Intended for research use only; not for human or veterinary applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.