Learn More
Cole-Parmer Cardboard Freezer Box Divider, 5-3/8" x 5-3/8" x 2-3/4", 36-Place, for 15 mL Tubes; 12/pk
Supplier:  Cole-Parmer 675579
Encompass_Preferred
Snug-fitting lid prevents accidental separation if dropped. Use in mechanical freezers or liquid nitrogen tanks to –196°C. Constructed of sturdy cardboard with moisture-repellent coating. Designed with small holes on the bottom to allow for drainage. Purchase cardboard freezer box separately to customize your storage needs.
Catalog No. NC1079838