Learn More
ABclonal Technology ARID1B Rabbit pAb
SDP
Supplier:  ABclonal Technology A154881000UL
Encompass_Preferred
This locus encodes an AT-rich DNA interacting domain-containing protein. The encoded protein is a component of the SWI/SNF chromatin remodeling complex and may play a role in cell-cycle activation. The protein encoded by this locus is similar to AT-rich interactive domain-containing protein 1A. These two proteins function as alternative, mutually exclusive ARID-subunits of the SWI/SNF complex. The associated complexes play opposing roles. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-07242