Learn More
Medchemexpress LLC Aripiprazole-d8 (OPC-14597-d8) | 1089115-06-9 | 98.35% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY14546S1MG
Encompass_Preferred
Aripiprazole-d8 (OPC-14597-d8) is a deuterium-labeled form of Aripiprazole (HY-14546). It acts as an atypical antipsychotic, functioning as a potent and high-affinity dopamine D2 receptor partial agonist. This compound also exhibits inverse agonist actions at 5-HT2B and 5-HT2A receptors, along with partial agonist effects at 5-HT1A, 5-HT2C, D3, and D4 receptors. It is suitable for research into conditions such as schizophrenia and COVID-19.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Acts as a potent and high-affinity dopamine D2 receptor partial agonist
  • Exhibits inverse agonist actions at 5-HT2B and 5-HT2A receptors
  • Displays partial agonist actions at 5-HT1A, 5-HT2C, D3, and D4 receptors
  • Suitable for schizophrenia research
  • Suitable for COVID-19 research
Catalog No. 50-003-55394