Learn More
Cayman Chemical ArIpIprazole N-oxIde 25mg
SDP
Supplier:  Cayman Chemical 3529625 MG
Encompass_Preferred
A metabolite of aripiprazole; a potential impurity found in commercial preparations of aripiprazole
Catalog No. 50-291-3498