Learn More
Medchemexpress LLC HY-N2013 1mg Medchemexpress, Aristolactam I CAS:13395-02-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN20131MG
Encompass_Preferred
Medchemexpress, HY-N2013 1mg Aristolactam I CAS:13395-02-3 Aristololactam I (AL-I), is the main metabolite of aristolochic acid I (AA-I), participates in the processes that lead to renal damage.Aristololactam I (AL-I) directly injures renal proximal tubule cells, the cytotoxic potency of AL-I is higher than that of AA-I and that the cytotoxic effects of these molecules are mediated through the induction of apoptosis in a caspase-3-dependent pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0357