Learn More
Medchemexpress LLC Arl67156 triethylamine | 00-00-0 | 99.9% | 1154.23 | C15H24Br2N5O12P3.(4.3C6H15N) | 1 MG
SDP
Supplier:  Medchemexpress LLC HY103265D1MG
Encompass_Preferred
ARL67156 triethylamine is a small-molecule NTPDase (ecto-ATPase) inhibitor supplied as a solid research reagent for biochemical and pharmacological studies of extracellular nucleotide metabolism and purinergic signaling.
  • Solid formulation suitable for storage and handling.
  • High purity for research applications.
  • Available as a 1 mg solid quantity.
  • Used as a competitive inhibitor of NTPDase (CD39), NTPDase3, and NPP1.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-46957