Learn More
Medchemexpress LLC Arl67156 triethylamine | 00-00-0 | MFCD12910436 | 99.9% | 1154.23 g/mol | C15H24Br2N5O12P3.(4.3C6H15N) | 100 MG
SDP
Supplier:  Medchemexpress LLC HY103265D100MG
Encompass_Preferred
ARL67156 triethylamine is a small-molecule inhibitor of ecto-ATPase enzymes (NTPDase family) used in cellular and animal research to study purinergic signaling and related disease models. It is supplied in solid and solution formats and is intended for research use only.
  • Selective inhibitor of NTPDase1, NTPDase3, and NPP1.
  • Available as solid (including 100 mg) and 10 mM solution formats.
  • High purity (approx. 99.9%) suitable for biochemical assays.
  • Storage: solid at 4°C; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
  • Used in studies of purinergic signaling and disease models such as aortic valve calcification and asthma.
Catalog No. 50-002-46300