Learn More
Medchemexpress LLC ARL67156 triethylamine | MFCD12910436 | 99.9% | 1154.23 g/mol | C15H24Br2N5O12P3 · (4.3 C6H15N) | 5 MG
SDP
Supplier:  Medchemexpress LLC HY103265D5MG
Encompass_Preferred
Triethylammonium salt of ARL67156, a small-molecule inhibitor of ecto-ATPases (NTPDases) including CD39 and CD73, supplied for research use. The solid is high purity (99.86%) and is provided in small quantities suitable for biochemical assays; recommended storage is 4°C for the solid and -80°C (6 months)/-20°C (1 month) for solutions.
Catalog No. 50-002-46005