Learn More
Medchemexpress LLC Arl67156 triethylamine | MFCD12910436 | 99.9% | 1154.23 g/mol | C15H24Br2N5O12P3.(4.3C6H15N) | 10 MG
SDP
Supplier:  Medchemexpress LLC HY103265D10MG
Encompass_Preferred
ARL67156 triethylamine is a research-grade small-molecule inhibitor of NTPDase enzymes supplied as a high-purity solid and offered in multiple small-package sizes and ready-to-use solution formats.
  • Inhibits NTPDase enzymes (CD39, NTPDase3, NPP1) in biochemical and cellular assays.
  • High purity solid suitable for analytical and preclinical research.
  • Available in small mg quantities and as 10 mM solutions for convenient use.
  • Store sealed at 4°C; in solvent, store at -80°C (6 months) or -20°C (1 month) per handling guidance.
  • Intended for research use only; not for human or clinical applications.
Catalog No. 50-002-45969