Learn More
Medchemexpress LLC ARMET/MANF Human H 100ug
SDP
Supplier:  Medchemexpress LLC HYP71940A100UG
Encompass_Preferred
Recombinant human ARMET/MANF protein with an N-terminal 6xHis tag, expressed in Escherichia coli and matching UniProt accession P55145 (L25-L182). Supplied for research use and characterized by certificate of analysis with a reported biological activity of 0.8862 μg/mL, it is primarily used in studies of neuronal survival, endoplasmic reticulum stress, and modulation of inhibitory neurotransmission.
  • Recombinant human ARMET/MANF with N-terminal 6xHis tag.
  • Expressed in Escherichia coli for scalable production.
  • Corresponds to UniProt P55145 residues L25-L182.
  • Characterized biological activity of 0.8862 μg/mL (per COA).
  • Suitable for research on neuronal survival, ER stress, and neurotransmission.
Catalog No. 50-001-96408