Learn More
Analytical Sales & Services Armor C18 Analytical Column. 120Â, 3µm, 3cm x 4.6mm ID
Supplier:  Analytical Sales & Services ADV7329
Encompass
Armor C18 Analytical Column. 120Â, 3µm, 3cm x 4.6mm ID
Catalog No. 50-008-05752