Learn More
Medchemexpress LLC HY-110101 10mg Medchemexpress, ARN272 CAS:488793-85-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11010110MG
Encompass_Preferred
Medchemexpress, HY-110101 10mg ARN272 CAS:488793-85-7 ARN272 is an anandamide transport inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8746