Learn More
Medchemexpress LLC Aromadendrene | 489-39-4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN841110MG
Encompass_Preferred
Aromadendrene can be isolated from Eucalyptus globulus and exhibits antimicrobial activity. It has a molecular weight of 204.35 and a chemical formula of C15H24. Its appearance is a colorless to light yellow liquid with a density of 0.9166 g/cm3. It is classified under terpenoids, specifically sesquiterpenes.
  • Antimicrobial activity
  • High purity: 96.00%
Catalog No. 50-004-17414