Learn More
Cayman Chemical ArphamenIn BhemIsulfate 10mg
SDP
Supplier:  Cayman Chemical 1850610 MG
Encompass_Preferred
An aminopeptidase B inhibitor that is commonly used to characterize novel proteases
Catalog No. 50-296-6006