Learn More
Aviva Systems Biology Corp ARPM1 Antibody - N-terminal region (ARP60226_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60226_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ARPM1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ARPM1. Alternate names: MGC15664, ARPM1ApplicationWB. Species reactivity: Horse, Human, Rabbit
Catalog No. 50-286-979