Learn More
TARGETMOL CHEMICALS INC ARQ 531 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1432310MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. ARQ-531 is a potent and orally active BTK inhibitor with potential antineoplastic activity, with IC50s of 0.85 nM and 0.39 nM for WT-BTK and C481S-BTK, respectively. Purity 99.59%
Catalog No. 50-287-6008