Learn More
Selleck Chemical LLC ARQ 531 25mg 2095393-15-8
SDP
Supplier:  Selleck Chemical LLC S871125MG
Encompass_Preferred
ARQ 531 is an ATP-competitive tyrosine kinase inhibitor designed to target BTK with an IC50 of 0.85 nM. It also has a distinct kinase selectivity profile with strong inhibitory activity against several key oncogenic drivers from TEC, Trk and Src family kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1704088