Learn More
Medchemexpress LLC ARQ 621 | 1095253-39-6 | 98.1% | 552.43 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16062100MG
Encompass_Preferred
ARQ 621 is an allosteric, potent, and selective inhibitor of Eg5, a microtubule-based ATPase motor protein involved in cell division. It exhibits anti-tumor activity and acts as a kinesin inhibitor. Additionally, ARQ 621 is a click chemistry reagent containing an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Allosteric, potent, and selective Eg5 inhibitor
  • Exhibits anti-tumor activity
  • Acts as a kinesin inhibitor
  • Click chemistry reagent with an alkyne group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-95255