Learn More
Medchemexpress LLC ARQ 621 | 1095253-39-6 | ≥99.0% | 552.43 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16062
Encompass_Preferred
ARQ 621 is an allosteric, potent, and selective inhibitor of Eg5, a microtubule-based ATPase motor protein involved in cell division. It exhibits anti-tumor activity and is a kinesin inhibitor. Additionally, ARQ 621 is a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Allosteric, potent, and selective inhibitor of Eg5
  • Microtubule-based ATPase motor protein inhibitor
  • Involved in cell division
  • Anti-tumor activity
  • Kinesin inhibitor
  • Click chemistry reagent with an alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-95374