Learn More
Medchemexpress LLC ARQ 621 | 1095253-39-6 | ≥99.0% | 552.43 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY160625MG
Encompass_Preferred
ARQ 621 is an allosteric, potent, and selective inhibitor of Eg5, which is a microtubule-based ATPase motor protein involved in cell division. It exhibits anti-tumor activity and acts as a kinesin inhibitor. Additionally, ARQ 621 is a click chemistry reagent containing an Alkyne group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Allosteric, potent and selective inhibitor of Eg5
  • Exhibits anti-tumor activity
  • Functions as a kinesin inhibitor
  • Click chemistry reagent with an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups
Catalog No. 50-003-94856