Learn More
Medchemexpress LLC ARQ 621 | 1095253-39-6 | ≥99.0% | 552.43 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1606250MG
Encompass_Preferred
ARQ 621 is an allosteric, potent and selective inhibitor of Eg5, a microtubule-based ATPase motor protein involved in cell division. Anti-tumor activity. ARQ 621 is a kinesin inhibitor. ARQ 621 is a click chemistry reagent, it contains an Alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. For research use only. We do not sell to patients.
  • ARQ 621 is an allosteric, potent and selective inhibitor of Eg5, a microtubule-based ATPase motor protein involved in cell division. It shows anti-tumor activity.
  • It is a kinesin inhibitor.
  • It is a click chemistry reagent, containing an Alkyne group, and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
Catalog No. 50-003-95254