Learn More
Medchemexpress LLC ARQ 621 | 1095253-39-6 | MFCD25976786 | 99.0% | 552.43 g/mol | C28H24Cl2FN5O2 | 10 MG

Supplier: Medchemexpress LLC HY1606210MG
ARQ 621 is a small-molecule, allosteric inhibitor of the mitotic motor protein Eg5 (kinesin spindle protein), used in biochemical and cell-based research to evaluate antiproliferative and mitotic-disruption effects.
- Potent and selective Eg5 inhibitor with antitumor activity.
- Purity ≥99.0%.
- Molecular weight 552.43 g/mol.
- Chemical formula C28H24Cl2FN5O2.
- Solid, white to off-white powder; soluble in DMSO (110 mg/mL).
- Store protected from light at -20°C; in solution, store at -80°C for up to 6 months.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.