Learn More
Selleck Chemical LLC ARQ 621
SDP
Supplier:  Selleck Chemical LLC S73555MG
Encompass_Preferred
ARQ 621 is an allosteric and selective Eg5 mitotic motor protein inhibitor Phase 1
Catalog No. 50-000-04610