Learn More
Medchemexpress LLC Ars-1630 | 1698055-86-5 | 430.84 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYU00417100MG
Encompass_Preferred
ARS-1630 is a novel inhibitor of mutant K-ras G12C, whose properties are derived from patent WO 2015054572 A1. It is a less active enantiomer of ARS-1620 and is supplied as a white to yellow solid.
  • Inhibits mutant K-ras G12C
  • Less active enantiomer of ARS-1620
  • Available in solid form
  • White to yellow appearance
Catalog No. 50-004-49668