Learn More
ATCC ART; Ovary Epithelium; Human (Homo sapiens)
Supplier:  ATCC CRL3468
Encompass
ART is a cell line exhibiting epithelial morphology that was derived from an existing ovarian cell line from a 64-year-old female. It can be used to evaluate the infectious entry of polyomaviruses (BKPyV, JCPyV, SV40, MCPyV) and human papillomaviruses. Pathogenic variants of the JCPyV virus previously believed to be non-infectious can infect this cell line.
Catalog No. 50-238-4378