Learn More
Medchemexpress LLC HY-141520 5mg , ART558 CAS:2603528-97-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1415205MG
Encompass_Preferred
HY-141520 5mg ART558 CAS: 2603528-97-6 ART558 is a nanomolar potent, selective, low molecular weight, allosteric DNA polymerase activity of Polθ inhibitor (IC50=7.9 nM). ART558 can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8949