Learn More
Medchemexpress LLC ART812 | 2607138-82-7 | 99.9% | 461.79 | 25 MG

Supplier: Medchemexpress LLC 25MGHY139289
ART812 is an orally active DNA polymerase Polθ inhibitor with an IC50 value of 7.6 nM. It also has an IC50 value of 240 nM for cell-based microhomology-mediated end joining (MMEJ). The compound elicits Polθ inhibitor sensitivity in MDA-MB-436 SHLD2 knockout cells and shows significant tumor inhibition in rats with MDA-MB-436 BRCA1/SHLD2 defective tumors.
- Orally active DNA polymerase Polθ inhibitor
- IC50 value of 7.6 nM for Polθ
- IC50 value of 240 nM for cell-based microhomology-mediated end joining (MMEJ)
- Elicits Polθ inhibitor sensitivity in MDA-MB-436 SHLD2 knockout cells
- Shows significant tumor inhibition in rats with MDA-MB-436 BRCA1/SHLD2 defective tumors
- Purity of 99.9%
- Molecular weight of 461.79
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.